missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decon™ Contrex™ AP Powdered Labware Detergent
Ideal for cleaning labware, scientific apparatus, and process equipment
$114.50 - $677.00
Specifications
| Product Type | Detergent |
|---|---|
| Color-coded | No |
| Form | Powder |
| pH | 9.5 to 11 |
| Scent | Regular Scent |
Description
Ideal for cleaning labware, scientific apparatus, and process equipment
- Concentrated powder
- Formulated for use in manual, soak, or ultrasonic cleaning
- Free-rinsing, completely soluble, and disposable straight to drain
- Available in a user-friendly bottle which permits easy dispensing and eliminates caking
Specifications
| Detergent | |
| Powder | |
| Regular Scent | |
| White |
| No | |
| 9.5 to 11 | |
| Non-sterile | |
| For use in manual, soak or ultrasonic cleaning; Cleans labware, scientific apparatus and process equipment |