Learn More
Corning™ DeckWorks™ Tip Stations, Sterile
Designed for single and multichannel pipetting stations
Supplier: Corning™ 4143
Description
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip Âstations contain 960 tips, are available in extended-length 10 μL and traditional 200 μL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only. Applications include pre- and Âpost-PCR procedures, cell culture, and assays requiring large tip quantities. Corning DeckWorks tip stations are offered both sterile and nonsterile.
Specifications
| 0.1 to 10 μL | |
| Natural | |
| Multi-pack | |
| Yes | |
| Polypropylene |
| Sterile | |
| Round | |
| Universal Fit, Multichannel Pipetting Stations | |
| DeckWorks |