missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ DeckWorks™ Reload Systems Starter Kits
Contains one DeckWorks Reload System with standard tips and empty hinged racks
$131.89 - $189.35
Specifications
| Sterility | Non-sterile |
|---|---|
| For Use With (Application) | Designed to allow to maximize storage space and help reduce the production and waste of plastic materials |
| Format | Refill |
| Tip Style | Round |
| Packaging | Starter kit |
| Catalog Number | Mfr. No. | Volume (Metric) | Color | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Color | Price | Quantity | ||||
|
10-320-721
|
Corning™
4132 |
1 to 200 μL | Yellow |
Case of 960 Each for $150.03
|
|
||||
|
10-320-719
|
Corning™
C4130 |
0.1 to 10 μL | Natural |
Case of 960 Each for $131.89
|
|
||||
|
10-320-723
|
Corning™
4134 |
100 to 1000 μL | Natural |
Case of 768 Each for $189.35
|
|
||||
|
10-320-722
|
Corning™
4133 |
1 to 300 μL | Natural |
Case of 960 Each for $153.17
|
|
||||
|
10-320-720
|
Corning™
4131 |
1 to 200 μL | Natural |
Case of 960 Each for $131.89
|
|
||||
Description
DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks.
- 10 empty racks accompany 10 μL, 200 μL, and 300 μL volume tips.
- 8 empty racks accompany 1000 μL volume tips.
Specifications
| Non-sterile | |
| Refill | |
| Starter kit | |
| DeckWorks |
| Designed to allow to maximize storage space and help reduce the production and waste of plastic materials | |
| Round | |
| Yes | |
| Polypropylene |