missing translation for 'onlineSavingsMsg'
Learn More

Decanoyl Chloride 98.0+%, TCI America™

Catalog No. D0027500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0027500ML 500 mL
D002725ML 25 mL
D0027100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0027500ML Supplier TCI America Supplier No. D0027500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-13-0
Molecular Formula C10H19ClO
Molecular Weight (g/mol) 190.711
MDL Number MFCD00000771
InChI Key IPIVAXLHTVNRBS-UHFFFAOYSA-N
Synonym capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n
PubChem CID 66982
IUPAC Name decanoyl chloride
SMILES CCCCCCCCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 230°C
Quantity 500 mL
UN Number 3265
Formula Weight 190.71
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Decanoyl Chloride
EINECSNumber (2)-0632
TSCA Yes