missing translation for 'onlineSavingsMsg'
Learn More

Decanoyl chloride, 98%

Catalog No. AAA1948622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1948622 100 g
AAA1948636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1948622 Supplier Thermo Scientific Chemicals Supplier No. A1948622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-13-0 | C10H19ClO | 190.711 g/mol

Decanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 112-13-0
Molecular Formula C10H19ClO
Molecular Weight (g/mol) 190.711
MDL Number MFCD00000771
InChI Key IPIVAXLHTVNRBS-UHFFFAOYSA-N
Synonym capric acid chloride, n-decanoyl chloride, capric chloride, decanoic acid chloride, caprinoyl chloride, caprinic acid chloride, n-decanoic acid chloride, decanoyl chloride, acmc-1c5ty, ksc182e3n
PubChem CID 66982
IUPAC Name decanoyl chloride
SMILES CCCCCCCCCC(=O)Cl

Specifications

Melting Point -35°C
Density 0.925
Boiling Point 232°C
Flash Point 106°C (223°F)
Odor Irritating
Refractive Index 1.441
Quantity 100 g
UN Number UN3265
Beilstein 507055
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 190.71
Percent Purity 98%
Chemical Name or Material Decanoyl chloride
Show More Show Less
Hazard Category H290-H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 203-938-0
RTECSNumber HE0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only