missing translation for 'onlineSavingsMsg'
Learn More

Decanophenone 98.0+%, TCI America™

Catalog No. D002625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D002625G 25 g
D0026500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D002625G Supplier TCI America Supplier No. D002625G

Chemical Identifiers

CAS 6048-82-4
Molecular Formula C16H24O
Molecular Weight (g/mol) 232.37
MDL Number MFCD00009582
InChI Key QQXJNLYVPPBERR-UHFFFAOYSA-N
Synonym decanophenone, 1-decanone, 1-phenyl, n-decanophenone, capriphenone, ketone, nonyl phenyl, ketone, nonenyl phenyl, nonyl phenyl ketone, 1-phenyl-1-decanone, acmc-209mkb
PubChem CID 80148
IUPAC Name 1-phenyldecan-1-one
SMILES CCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 187°C
Quantity 25 g
Formula Weight 232.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Decanophenone