missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decanoic Acid 98.0+%, TCI America™
Supplier: TCI America D0017400G
Specifications
| Decanoic Acid | |
| 270°C | |
| C10H20O2 | |
| capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid | |
| CCCCCCCCCC(=O)O | |
| 172.268 | |
| CHEBI:30813 | |
| ≥98.0% (GC,T) | |
| Crystalline |
| 334-48-5 | |
| White | |
| MFCD00004441 | |
| GHVNFZFCNZKVNT-UHFFFAOYSA-N | |
| decanoic acid | |
| 2969 | |
| 172.27 | |
| 400 g |
Chemical Identifiers
| 334-48-5 | |
| 172.268 | |
| GHVNFZFCNZKVNT-UHFFFAOYSA-N | |
| 2969 | |
| decanoic acid |
| C10H20O2 | |
| MFCD00004441 | |
| capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid | |
| CHEBI:30813 | |
| CCCCCCCCCC(=O)O |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : HD9100000
TSCA : Yes