missing translation for 'onlineSavingsMsg'
Learn More

Decanoic Acid 98.0+%, TCI America™

Catalog No. D001725G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D001725G 25 g
D0017400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D001725G Supplier TCI America Supplier No. D001725G

Chemical Identifiers

CAS 334-48-5
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00004441
InChI Key GHVNFZFCNZKVNT-UHFFFAOYSA-N
Synonym capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
PubChem CID 2969
ChEBI CHEBI:30813
IUPAC Name decanoic acid
SMILES CCCCCCCCCC(=O)O

Specifications

Boiling Point 270°C
Color White
Formula Weight 172.27
Percent Purity ≥98.0% (GC,T)
Quantity 25 g
Physical Form Crystalline
Chemical Name or Material Decanoic Acid
EINECSNumber (2)-0608
RTECSNumber HD9100000
TSCA Yes