Learn More
Decanoic acid, 99%
CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
Supplier: Thermo Scientific Chemicals A147880I
Description
Decanoic acid is used in perfumes, resins, lubricants, greases, dyes, plastics, pharmaceuticals and food additives. It is involved in the preparation of wetting agents and sodium deconate, which is an active component in soap. It is also used as a plasticizer and as an absorption enhancer in guinea pig skin experiments. Further, it is employed in the generation of esters for artificial fruit flavors. In addition to this, it is used in agricultural chemicals, lubricant additives and surface active agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDecanoic acid is used in perfumes, resins, lubricants, greases, dyes, plastics, pharmaceuticals and food additives. It is involved in the preparation of wetting agents and sodium deconate, which is an active component in soap. It is also used as a plasticizer and as an absorption enhancer in guinea pig skin experiments. Further, it is employed in the generation of esters for artificial fruit flavors. In addition to this, it is used in agricultural chemicals, lubricant additives and surface active agents.
Solubility
Soluble in acetone, benzene, chloroform, carbon disulfide, ethyl ether, ethanol, dilute nitric acid and most organic solvents. Insoluble in water.
Notes
Incompatible with bases, reducing agents and oxidizing agents.
Specifications
| Decanoic acid | |
| 29°C to 33°C | |
| 268°C to 270°C | |
| C10H20O2 | |
| 1754556 | |
| capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid | |
| GHVNFZFCNZKVNT-UHFFFAOYSA-N | |
| decanoic acid | |
| 2969 | |
| 172.27 | |
| 0.893 | |
| 147°C (296°F) |
| 4 | |
| 334-48-5 | |
| 99% | |
| MFCD00004441 | |
| 14,1758 | |
| Soluble in acetone,benzene,chloroform,carbon disulfide,ethyl ether,ethanol,dilute nitric acid and most organic solvents. Insoluble in water. | |
| CCCCCCCCCC(=O)O | |
| 172.268 | |
| CHEBI:30813 | |
| 99% | |
| 5000 g |
Chemical Identifiers
| 334-48-5 | |
| 172.268 | |
| GHVNFZFCNZKVNT-UHFFFAOYSA-N | |
| 2969 | |
| decanoic acid |
| C10H20O2 | |
| MFCD00004441 | |
| capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid | |
| CHEBI:30813 | |
| CCCCCCCCCC(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 206-376-4
RTECSNumber : HD9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only