missing translation for 'onlineSavingsMsg'
Learn More

Decanoic acid, 99%, Thermo Scientific™

Catalog No. AC167270025
Change view
Click to view available options
2.5kg; Plastic drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC167270025 2.5kg; Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC167270025 Supplier Thermo Scientific Chemicals Supplier No. 167270025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 334-48-5
Assay Percent Range 99%
Molecular Formula C10H20O2
Linear Formula CH3(CH2)8CO2H
MDL Number MFCD00004441
Beilstein 02, II, 309
Merck Index 15, 1760
Synonym capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
InChI Key GHVNFZFCNZKVNT-UHFFFAOYSA-N
SMILES CCCCCCCCCC(=O)O
IUPAC Name decanoic acid
Molecular Weight (g/mol) 172.268
PubChem CID 2969
ChEBI CHEBI:30813
Formula Weight 172.27
Percent Purity 99%
Density 0.8900g/mL
Quantity 2.5kg
Flash Point 150°C
Infrared Spectrum Authentic
Packaging Plastic Drum
Specific Gravity 0.89
Chemical Name or Material Decanoic acid, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-376-4
RTECSNumber HD9100000
TSCA TSCA

RUO – Research Use Only