missing translation for 'onlineSavingsMsg'
Learn More

Decane 99.0+%, TCI America™

Catalog No. p-7125981
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D001125ML 25 mL
D0011500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D001125ML Supplier TCI America Supplier No. D001125ML
Only null left

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Melting Point -30°C
Color Colorless
Boiling Point 174°C
Quantity 25 mL
UN Number 2247
Formula Weight 142.29
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Decane
EINECSNumber (2)-0010
RTECSNumber HD6550000
TSCA Yes