missing translation for 'onlineSavingsMsg'
Learn More

Decamethylzirconocene Dichloride 95.0+%, TCI America™

Catalog No. D33071G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33071G Supplier TCI America Supplier No. D33071G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54039-38-2
Molecular Formula C20H30Cl2Zr-2
Molecular Weight (g/mol) 432.584
InChI Key OCFSLQKTBFSWPL-UHFFFAOYSA-L
Synonym decamethylzirconocene dichloride
PubChem CID 57369741
IUPAC Name dichlorozirconium;1,2,3,4,5-pentamethylcyclopenta-1,3-diene
SMILES C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.Cl[Zr]Cl

Specifications

Color Yellow
Quantity 1 g
UN Number 3261
Formula Weight 432.58
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Decamethylzirconocene Dichloride