missing translation for 'onlineSavingsMsg'
Learn More

Decamethyltetrasiloxane 97.0+%, TCI America™

Catalog No. D231425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D231425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D231425ML Supplier TCI America Supplier No. D231425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-62-8
Molecular Formula C10H30O3Si4
Molecular Weight (g/mol) 310.69
MDL Number MFCD00008263
InChI Key YFCGDEUVHLPRCZ-UHFFFAOYSA-N
PubChem CID 8852
IUPAC Name 2,2,4,4,6,6,8,8-octamethyl-3,5,7-trioxa-2,4,6,8-tetrasilanonane
SMILES C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 194°C
Quantity 25 mL
Formula Weight 310.69
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Decamethyltetrasiloxane
EINECSNumber (7)-0476
TSCA Yes