missing translation for 'onlineSavingsMsg'
Learn More

Decamethylcyclopentasiloxane 99.0+%, TCI America™

Catalog No. D1890500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1890500ML 500 mL
D189025ML 25 mL
D1890100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1890500ML Supplier TCI America Supplier No. D1890500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 541-02-6
Molecular Formula C10H30O5Si5
Molecular Weight (g/mol) 370.77
MDL Number MFCD00046966
InChI Key XMSXQFUHVRWGNA-UHFFFAOYSA-N
Synonym decamethylcyclopentasiloxane, cyclopentasiloxane, decamethyl, cyclomethicone 5, dimethylsiloxane pentamer, dow corning 345 fluid, nuc silicone vs 7158, dow corning 345, silicon sf 1202, cyclic dimethylsiloxane pentamer, dekamethylcyklopentasiloxan
PubChem CID 10913
IUPAC Name 2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
SMILES C[Si]1(O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)C

Specifications

Melting Point -38°C
Color Colorless
Boiling Point 210°C
Quantity 500 mL
Formula Weight 370.77
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Decamethylcyclopentasiloxane
EINECSNumber (7)-0475
RTECSNumber GY5945200
TSCA Yes