missing translation for 'onlineSavingsMsg'
Learn More

Decamethylcyclopentasiloxane, 97%

Catalog No. AC454582500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454582500 250 g
AC454580500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454582500 Supplier Thermo Scientific Chemicals Supplier No. 454582500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 541-02-6 | C10H30O5Si5 | 370.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 541-02-6
Molecular Formula C10H30O5Si5
Molecular Weight (g/mol) 370.77
InChI Key XMSXQFUHVRWGNA-UHFFFAOYSA-N
Synonym decamethylcyclopentasiloxane, cyclopentasiloxane, decamethyl, cyclomethicone 5, dimethylsiloxane pentamer, dow corning 345 fluid, nuc silicone vs 7158, dow corning 345, silicon sf 1202, cyclic dimethylsiloxane pentamer, dekamethylcyklopentasiloxan
PubChem CID 10913
IUPAC Name 2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
SMILES C[Si]1(O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)C

Specifications

Density 0.9590g/mL
Boiling Point 101.0°C (20.0 mmHg)
Flash Point 76°C
Assay Percent Range 96% min. (GC)
Quantity 250 g
Specific Gravity 0.959
Merck Index 15,2852
Solubility Information Solubility in water: insoluble
Viscosity 4 mPa.s (20°C)
Formula Weight 370.77
Percent Purity 97%
Chemical Name or Material Decamethylcyclopentasiloxane
Show More Show Less
EINECSNumber 208-764-9

RUO – Research Use Only