missing translation for 'onlineSavingsMsg'
Learn More

Decamethonium Iodide, TCI America™

Catalog No. d13155g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D13155G Supplier TCI America Supplier No. D13155G
Only null left

Chemical Identifiers

CAS 1420-40-2
Molecular Formula C16H38I2N2
Molecular Weight (g/mol) 512.303
MDL Number MFCD00031553
InChI Key ARMLJSXXXFXSLQ-UHFFFAOYSA-L
Synonym Decamethylenebis(trimethylammonium Iodide), Decane-1,10-bis(trimethylammonium Iodide)
PubChem CID 74014
IUPAC Name trimethyl-[10-(trimethylazaniumyl)decyl]azanium;diiodide
SMILES C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C.[I-].[I-]

Specifications

Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 512.30
Physical Form Crystalline Powder
Chemical Name or Material Decamethonium Iodide
EINECSNumber (9)-1915
RTECSNumber BP6300000
TSCA No