missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decamethonium Bromide 98.0+%, TCI America™
$180.81 - $544.30
Chemical Identifiers
| CAS | 541-22-0 |
|---|---|
| Molecular Formula | C16H38Br2N2 |
| Molecular Weight (g/mol) | 418.302 |
| MDL Number | MFCD00011779 |
| InChI Key | HLXQFVXURMXRPU-UHFFFAOYSA-L |
| Synonym | decamethonium bromide, decamethonium dibromide, decacuran, syncurine, decamethylene bis trimethylammonium bromide, dekamethoniumbromid czech, decametonio bromuro dcit, unii-55c6rk944k, decamethylenebis trimethylammonium bromide |
| PubChem CID | 10921 |
| IUPAC Name | trimethyl-[10-(trimethylazaniumyl)decyl]azanium;dibromide |
| SMILES | C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C.[Br-].[Br-] |
Chemical Identifiers
| 541-22-0 | |
| 418.302 | |
| HLXQFVXURMXRPU-UHFFFAOYSA-L | |
| 10921 | |
| C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C.[Br-].[Br-] |
| C16H38Br2N2 | |
| MFCD00011779 | |
| decamethonium bromide, decamethonium dibromide, decacuran, syncurine, decamethylene bis trimethylammonium bromide, dekamethoniumbromid czech, decametonio bromuro dcit, unii-55c6rk944k, decamethylenebis trimethylammonium bromide | |
| trimethyl-[10-(trimethylazaniumyl)decyl]azanium;dibromide |
Specifications
| 541-22-0 | |
| C16H38Br2N2 | |
| 5 g | |
| decamethonium bromide, decamethonium dibromide, decacuran, syncurine, decamethylene bis trimethylammonium bromide, dekamethoniumbromid czech, decametonio bromuro dcit, unii-55c6rk944k, decamethylenebis trimethylammonium bromide | |
| C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C.[Br-].[Br-] | |
| 418.302 | |
| 418.30 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00011779 | |
| 2811 | |
| HLXQFVXURMXRPU-UHFFFAOYSA-L | |
| trimethyl-[10-(trimethylazaniumyl)decyl]azanium;dibromide | |
| 10921 | |
| ≥98.0% (T) | |
| Decamethonium Bromide |
Safety and Handling
EINECSNumber : (2)-0175
RTECSNumber : BP5950000
TSCA : Yes