Learn More
StatLab DecalStat Decalcifier
Catalog No. STL12121
Change view
Click to view available options
Product Type:
Decalcifying Solution
Catalog No. Product Type
STL12121 Decalcifying Solution
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. STL12121 Supplier StatLab Supplier No. 12121
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Harmful if swallowed, in contact with skin, or if inhaled
Decalcifies the average bone marrow biopsy in less than one hour.
  • Can be used on all types of calcified tissue and is especially effective on bone marrow samples
  • Cellular and nuclear details remain crisp and clear
  • 1 gallon, 4 per case
Specifications
Product Type Decalcifying Solution
Quantity Each
Solution Type Decalcifier
Form Solution