missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene (Laboratory), Fisher Chemical™

$83.38 - $83.38

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00004130
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
D0-100
SDS View Documents
Fisher Chemical
BPD0100
100 mL Glass Bottle
Each for $83.38
Case of 6 Each for $500.28
Only null left
 
Description

Description

Specifications

Chemical Identifiers

91-17-8
138.254
NNBZCPXTIHJBJL-UHFFFAOYSA-N
7044
1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
C10H18
MFCD00004130
decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
CHEBI:38853
C1CCC2CCCCC2C1

Specifications

91-17-8
Colorless
7
Glass Bottle
MFCD00004130
1147
NNBZCPXTIHJBJL-UHFFFAOYSA-N
1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
4.8g/mol
CHEBI:38853
1.3mbar at 22°C
Liquid
-31°C
0.890g/cm³
187°C
C10H18
100 mL
decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
C1CCC2CCCCC2C1
138.254
7044
138.25
Laboratory
Decahydronaphthalene
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Causes eye, skin, and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Handle under an inert atmosphere. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:2
Instability:1

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
Documents

Documents