missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene (Laboratory), Fisher Chemical™

Catalog No. D0100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D0100 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0100 Supplier Fisher BioReagents Supplier No. BPD0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00004130
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Melting Point -31°C
Color Colorless
Density 0.890g/cm³
pH 7
Boiling Point 187°C
Packaging Glass Bottle
Quantity 100 mL
UN Number 1147
Vapor Density 4.8g/mol
Vapor Pressure 1.3mbar at 22°C
Formula Weight 138.25
Grade Laboratory
Physical Form Liquid
Chemical Name or Material Decahydronaphthalene
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Causes eye, skin, and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Handle under an inert atmosphere. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:2
Instability:1
DOTInformation DOT Class 3, : Flammable Liquid