missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. D000825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D000825ML 25 mL
D0008500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D000825ML Supplier TCI America Supplier No. D000825ML

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00004130
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Color Colorless
Boiling Point 190°C
Quantity 25 mL
UN Number 1147
Formula Weight 138.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Decahydronaphthalene (cis- and trans- mixture)
EINECSNumber (4)-0575
RTECSNumber QJ3150000
TSCA Yes