missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene, cis + trans, 98%

Catalog No. aaa13883ap
Change view
Click to view available options
Quantity:
10,000 mL
100 mL
500 mL
2500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13883AP 500 mL
AAA13883AE 100 mL
AAA138830F 2500 mL
AAA138830D 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA13883AP Supplier Thermo Scientific Chemicals Supplier No. A13883AP

CAS: 91-17-8 | C10H18 | 138.254 g/mol

Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.

Solubility
Miscible with acetone, benzene, ether, methanol, aniline, decalin, chloroform, butanol and ethanol. Immiscible with water.

Notes
Store in cool place. Air, light and heat sensitive. Heat and light accelerate peroxide formation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00004130
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Melting Point <-45°C
Density 0.88
Boiling Point 189°C to 191°C
Flash Point 57°C (134°F)
Odor Mild
Refractive Index 1.474
Quantity 500 mL
UN Number UN1147
Beilstein 878165
Merck Index 14,2846
Solubility Information Miscible with acetone,benzene,ether,methanol,aniline,decalin,chloroform,butanol and ethanol. Immiscible with water.
Formula Weight 138.25
Percent Purity 98%
Chemical Name or Material Decahydronaphthalene, cis + trans
Show More Show Less
Hazard Category H226-H304-H314-H331-H335
Hazard Statement GHS H Statement
H331-H226-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DECAHYDRONAPHTHALENE
EINECSNumber 202-046-9
RTECSNumber QJ3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only