Learn More
Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™
CAS: 91-17-8 | C10H18 | 138.25 g/mol
$65.41 - $65.41
Chemical Identifiers
| CAS | 91-17-8 |
|---|---|
| Molecular Formula | C10H18 |
| Molecular Weight (g/mol) | 138.25 |
| InChI Key | NNBZCPXTIHJBJL-UHFFFAOYSA-N |
| Synonym | decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane |
| PubChem CID | 7044 |
| ChEBI | CHEBI:38853 |
| IUPAC Name | 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene |
| SMILES | C1CCC2CCCCC2C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC406171000
|
Thermo Scientific Chemicals
406171000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $65.41
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 91-17-8 | |
| 138.25 | |
| decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane | |
| CHEBI:38853 | |
| C1CCC2CCCCC2C1 |
| C10H18 | |
| NNBZCPXTIHJBJL-UHFFFAOYSA-N | |
| 7044 | |
| 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene |
Specifications
| 91-17-8 | |
| 0.8900g/mL | |
| 58°C | |
| 98.5% min. (mixture of cis and trans) (GC) | |
| C10H18 | |
| C10H18 | |
| 05,III,245 | |
| 15,285 | |
| Solubility in water: slightly soluble (6mg/l at 20°C). Other solubilities: very soluble in acetone,benzene,ether and,ethanol | |
| C1CCC2CCCCC2C1 | |
| 138.25 | |
| 3 mPa.s (20°C) | |
| 138.25 | |
| Liquid |
| -31.0°C | |
| 187.0°C | |
| Authentic | |
| AcroSeal™ Glass Bottle | |
| 1.4740 to 1.4760 | |
| 100 mL | |
| 0.89 | |
| decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane | |
| NNBZCPXTIHJBJL-UHFFFAOYSA-N | |
| 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene | |
| 7044 | |
| CHEBI:38853 | |
| 99% | |
| Decahydronaphthalene |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 202-046-9
RUO – Research Use Only