missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decahydronaphthalene, 98%, mixture of cis and trans
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -31.0°C |
| Density | 0.8900g/mL |
| Boiling Point | 187.0°C |
| Flash Point | 58°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4740 to 1.4760 |
| Linear Formula | C10H18 |
| Quantity | 1 L |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Toxic if inhaled. May be fatal if swallowed and enters airways. Very toxic to aquatic life with long lasting effects. Flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye p |
| EINECSNumber | 202-046-9 |
RUO – Research Use Only