missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decahydroisoquinoline (cis- and trans- mixture, predominantly cis-isomer) 98.0+%, TCI America™
$328.08 - $986.08
Chemical Identifiers
| CAS | 6329-61-9 |
|---|---|
| Molecular Formula | C9H17N |
| Molecular Weight (g/mol) | 139.24 |
| MDL Number | MFCD00012096 |
| InChI Key | NENLYAQPNATJSU-UHFFFAOYNA-N |
| PubChem CID | 97812 |
| IUPAC Name | decahydroisoquinoline |
| SMILES | C1CCC2CNCCC2C1 |
Chemical Identifiers
| 6329-61-9 | |
| 139.24 | |
| NENLYAQPNATJSU-UHFFFAOYNA-N | |
| decahydroisoquinoline |
| C9H17N | |
| MFCD00012096 | |
| 97812 | |
| C1CCC2CNCCC2C1 |
Specifications
| 6329-61-9 | |
| 80°C | |
| MFCD00012096 | |
| NENLYAQPNATJSU-UHFFFAOYNA-N | |
| decahydroisoquinoline | |
| 97812 | |
| ≥98.0% (GC) | |
| Decahydroisoquinoline (cis- and trans- mixture, predominantly cis-isomer) |
| Yellow | |
| C9H17N | |
| 5 mL | |
| C1CCC2CNCCC2C1 | |
| 139.24 | |
| 139.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-6524
TSCA : No