missing translation for 'onlineSavingsMsg'
Learn More

Decacarbonyldirhenium, 96%, Thermo Scientific Chemicals

Catalog No. p-7080298
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA6210803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA6210803 Supplier Thermo Scientific Chemicals Supplier No. 06210803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14285-68-8 | Molecular Formula: C10O10Re2 | Molecular Weight: 652.51 g/mol

Decacarbonyldirhenium is a key starting material used for the synthesis of other rhenium carbonyl complexes. It plays an important role in metathesis, reforming and hydrogenation reactions. It is used as a catalyst to promote silation of alcohols and in the preparion of silyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14285-68-8
Molecular Formula C10O10Re2
Molecular Weight (g/mol) 652.51
MDL Number MFCD00011198
InChI Key ZIZHEHXAMPQGEK-UHFFFAOYSA-N
Synonym carbon monoxide; rhenium 2+, decacarbonyldirhenium 250mg
PubChem CID 498777
SMILES [O]#C[Re](C#[O])(C#[O])(C#[O])(C#[O])[Re](C#[O])(C#[O])(C#[O])(C#[O])C#[O]

Specifications

Melting Point 170°C (decomposition)
Color White
Density 2.87 g/mL
Linear Formula Re2(CO)10
Quantity 1 g
UN Number UN3466
IUPAC Name carbon monoxide;rhenium
Formula Weight 652.52
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Decacarbonyldirhenium
Show More Show Less
Hazard Category H332
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P271-P304+P340-P312
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber 238-202-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only