missing translation for 'onlineSavingsMsg'
Learn More

DEAE-Dextran (Average mol wt. 500,000), MilliporeSigma™ Supelco™

Catalog No. 111015843
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015843 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015843 Supplier Merck Emd Millipore Supplier No. 935561G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DEAE-Dextran is used as mediator in DNA transfection procedures. It is considered to be simple and effective.

Chemical Identifiers

Synonym Diethylaminoethyl-Dextran

Specifications

CAS 9015-73-0
Quantity 1 g
RTECSNumber HH9250000