Learn More
Waters Limited Deactivated Clear Glass 12 x 32 mm Snap Neck Qsert Vial, with Cap and PTFE/Silicone Septum, 300 L Volume, 100/pk
Catalog No. 50782533
Encompass
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.
Catalog No. 50782533 Supplier Waters Limited Supplier No. 186001124DV
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Deactivated vials have a silanized surface to prevent adsorption of highly polar molecules.