missing translation for 'onlineSavingsMsg'
Learn More

Chromium Acetate, Basic, Powder, Purified, Spectrum™ Chemical

Catalog No. 18609748
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609748 45 kg Fiber Drum
18609744 125 g Amber Glass Bottle
18609745 500 g Amber Glass Bottle
18609746 2.5 kg Poly Pail
18609747 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609748 Supplier Spectrum Chemical Mfg Cor Supplier No. C123645KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39430-51-8

Chemical Identifiers

CAS 39430-51-8

Specifications

CAS Max % 1
Physical Form Powder
Quantity 45 kg
Grade Purified
Packaging Fiber Drum
Chemical Name or Material Chromium Acetate, Basic, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.