missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Absolute, Reagent, ACS, 99.8%, Spectrum™ Chemical

Catalog No. 18604360
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18604360 200 L Drum
18604352 100 mL Amber Glass Bottle
18604353 500 mL Amber Glass Bottle
18604355 1 L Amber Glass Bottle
18604354 6 x 500 mL Amber Glass Bottle
18604356 4 L Amber Glass Bottle
18604357 6 x 1 L Amber Glass Bottle
18604358 4 x 4 L Amber Glass Bottle
18604359 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Numéro de catalogue. 18604360 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur M1240200LTBL
Get Up to $300 in Additional Chemicals
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

M1240, 67-56-1, CH3OH

Spectrum™ Chemical Methanol, Absolute, Reagent, ACS , is also called methyl alcohol or 'wood alcohol' it was one of the by products of distillation of wood. As the simplest alcohol around, it is light, flammable, colorless liquid with a strong and distinctive odor. It has many uses including acting as an antifreeze, (due to it's polar liquid properties), solvent, fuel as well as a denaturant for ethanol. As an ACS grade Reagent, Spectrum Chemical manufactured methanol is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Identifiants chimiques

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Spécifications

CAS Max % 100%
Quantity 200 L
Packaging Drum
Residue after Evaporation 0.001%
UN Number UN1230
Boiling Point 64°C to 65°C
Percent Purity 99.8%
Grade Reagent
Chemical Name or Material Methanol