missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ DBI-Sala™ Sealed Self-Retracting Lifeline: Carrying Cases
Description
For Self-Retracting Lifelines, see cat. no. 18716.
Specifications
Specifications
| Color | Dark Blue with Yellow Stripes |
| Product Type | Carrying Case |
| Dimensions (L x W x H) | 22.5 x 14 x 12 in. |
| For Use With (Equipment) | 85 ft. Self retracting lifeline |