missing translation for 'onlineSavingsMsg'
Learn More

3M™ DBI-Sala™ NanoLok™ Quick Connect Self Retracting Lifeline

Catalog No. 19070804
Change view
Click to view available options
Product Type:
Quick Connect Self Retracting Lifeline - Web
Twin-Leg Quick Connect Self Retracting Lifeline - Web
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19070804 Quick Connect Self Retracting Lifeline - Web
19070805 Twin-Leg Quick Connect Self Retracting Lifeline - Web
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19070804 Supplier 3M™ Supplier No. 70007443669
Only null left

3M Company DBI-SALA™ Nano-Lok™ Quick Connect Self Retracting Lifeline is a fiber and polyester web and aluminum rebar hook(s), quick connector for harness mounting.

  • Built-in connector for SRL direct attachment to harness
  • 6ft. (1.8m) lifelines with 3/4in. (19mm) Dyneema™ fiber and polyester web
  • Aluminum self-locking rebar hook on each lifeline
  • Extremely compact, lightweight design
  • Tough and impact resistant thermoplastic housing
  • Swiveling anchorage loops
  • Quick activating speed sensing brake systems
  • High capacity external energy absorbers and impact indicators
  • i-Safe™ equipped
  • Wide variety of configurations available

Certifications

ANSI A10.32, OSHA 1926.502, OSHA 1910.66, ANSI Z359.1, ANSI Z359.14

Compliance

Meets ANSI™ Z359.14-2012 requirements.

Specifications

Material Nylon Thermoplastic
Certifications/Compliance ANSI A10.32, OSHA 1926.502, OSHA 1910.66, ANSI Z359.1, ANSI Z359.14
Length (English) 6 ft.
For Use With (Equipment) Harnesses
Weight (English) 2.9 lb.
Product Type Quick Connect Self Retracting Lifeline - Web
Load Bearing Capacity (English) 420lb.
Load Bearing Capacity (Metric) 190 kg