Learn More
3M™ DBI-Sala™ Crossover Full-Body Harnesses
Lightweight polyester for all-day wear
Supplier: 3M™ 7012815449
Description
- Feature patented crossover design with adjustable front D-ring, used for positioning or ladder climbing
- Back D-ring is used for fall arrest or restraint
- Friction buckles on shoulders and mating pass-through adjuster buckles on lower shoulder straps and leg straps
- Side D-rings for additional restraint or positioning options
- Suitable for fall arrest, restraint, or positioning
Compliance
Meet ANSI Z359.1-1992, ANSI A10.14-1991, and OSHA requirements.
Specifications
| Small, Medium, Large | |
| Lightweight Polyester | |
| Side |