Learn More
3M™ DBI-Sala™ Construction Cross-over Style Harnesses
Catalog No. 19809216
Change view
missing translation for 'orderingAttributeHoverText'
Size:
Large
Medium
Small
X-Large
Catalog No. Size
19809216 X-Large
19054993 Small
19054994 Medium
19054995 Large
4 options
Catalog No. 19809216
Supplier: 3M™ 7012798774
Only null left
Construction harness with parachute and pass-through buckles
  • Material: Polyester
  • Cross-over style
  • No-tangle functionality
  • Front and back D-rings
  • Tongue buckle body belt w/foam back pad
  • Pass through legs
  • Belt and hip pad with side D-rings
  • Available in a variety of sizes
Specifications
Material Polyester Webbing
D-Ring Location Back, Side
Style Cross-over
Size X-Large