missing translation for 'onlineSavingsMsg'
Learn More

3M™ DBI-SALA™ Retrieval Wristlets

Catalog No. 19809185
Change view
Click to view available options
Includes:
Center O Ring, Wrist Loop On Each End
Wristlets Attached to a Y-lanyard; with Center D-ring
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19809185 Center O Ring, Wrist Loop On Each End
19809184 Wristlets Attached to a Y-lanyard; with Center D-ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19809185 Supplier 3M™ Supplier No. 7012815085

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Retrieval wristlets for confined space rescue.

  • Attaches to victims wrists
  • Ideal for narrow extractions
  • Integral D-ring connection point for winch/SRL
  • Durable polyester construction
  • Compact and lightweight design

Specifications

Color Yellow
Material Nylon
Product Type Wristlet
Certifications/Compliance OSHA, ANSI
For Use With (Equipment) Extrication lifeline
Includes Center O Ring, Wrist Loop On Each End