missing translation for 'onlineSavingsMsg'
Learn More

DBD-NCS [=4-(N,N-Dimethylaminosulfonyl)-7-isothiocyanato-2,1,3-benzoxadiazole], TCI America™

Catalog No. A5575100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5575100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5575100MG Supplier TCI America Supplier No. A5575100MG

[for HPLC Labeling and Edman Degradation]

Chemical Identifiers

CAS 147611-81-2
Molecular Formula C9H8N4O3S2
Molecular Weight (g/mol) 284.308
MDL Number MFCD03844763
InChI Key SUOZAUUDBKHLHQ-UHFFFAOYSA-N
Synonym 4-(N,N-Dimethylaminosulfonyl)-7-isothiocyanatobenzofurazan, 4-(N,N-Dimethylaminosulfonyl)-7-isothiocyanato-2,1,3-benzoxadiazole
PubChem CID 127043
IUPAC Name 7-isothiocyanato-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
SMILES CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)N=C=S

Specifications

Melting Point 126°C
Color Yellow
Quantity 100 mg
Formula Weight 284.31
Physical Form Crystalline Powder
Chemical Name or Material DBD-NCS [=4-(N,N-Dimethylaminosulfonyl)-7-isothiocyanato-2,1,3-benzoxadiazole] [for HPLC Labe
TSCA No
Recommended Storage Refrigerator