missing translation for 'onlineSavingsMsg'
Learn More

DBD-H 98.0+%, TCI America™

Catalog No. A5556100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5556100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5556100MG Supplier TCI America Supplier No. A5556100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 131467-86-2
Molecular Formula C8H11N5O3S
Molecular Weight (g/mol) 257.268
MDL Number MFCD00191376
InChI Key VPDDJYGQJSTHTA-UHFFFAOYSA-N
Synonym 4-(N,N-Dimethylaminosulfonyl)-7-hydrazinobenzofurazan, 4-(N,N-Dimethylaminosulfonyl)-7-hydrazino-2,1,3-benzoxadiazole
PubChem CID 131382
IUPAC Name 7-hydrazinyl-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
SMILES CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)NN

Specifications

Quantity 100 mg
UN Number 1325
Formula Weight 257.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material DBD-H [=4-(N,N-Dimethylaminosulfonyl)-7-hydrazino-2,1,3-benzoxadiazole] [for HPLC Labeling]
TSCA No
Recommended Storage Freezer