missing translation for 'onlineSavingsMsg'
Learn More

DBD-F 98.0+%, TCI America™

Catalog No. A5595100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5595100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5595100MG Supplier TCI America Supplier No. A5595100MG

Chemical Identifiers

CAS 98358-90-8
Molecular Formula C8H8FN3O3S
Molecular Weight (g/mol) 245.23
MDL Number MFCD00143356
InChI Key YZWANFXENNWOOR-UHFFFAOYSA-N
Synonym 4-(N,N-Dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole, 7-Fluoro-4-(N,N-dimethylaminosulfonyl)benzofurazan, 7-Fluoro-4-(N,N-dimethylsulfamoyl)-2,1,3-benzoxadiazole
PubChem CID 126917
IUPAC Name 7-fluoro-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
SMILES CN(C)S(=O)(=O)C1=CC=C(F)C2=NON=C12

Specifications

Melting Point 128°C
Color Yellow
Quantity 100 mg
Formula Weight 245.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material DBD-F [=4-(N,N-Dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] [for HPLC Labeling]
TSCA No
Recommended Storage Refrigerator