Learn More
Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers
Catalog No. 11676201
Change view
Click to view available options
Type:
RS-232 Output
RS-232 Output, Factory Installed
RS-485 Output
RS-485 Output, Factory Installed
Voltage:
0/1 V
0/5 V
4-20mA
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Voltage
11676201 RS-232 Output 0/5 V
11676200 RS-232 Output 0/1 V
11676202 RS-485 Output 4-20mA
11676203 RS-485 Output 0/1 V
11676204 RS-485 Output 0/5 V
13990503 RS-232 Output, Factory Installed 0/1 V
13990504 RS-485 Output, Factory Installed 0/1 V
13990505 RS-485 Output, Factory Installed 0/5 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 11676201 Supplier Thermo Scientific™ Supplier No. 195896
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:
  • RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
  • RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
Specifications
Type RS-232 Output
For Use With (Equipment) Forma™ 8600, HERAfreeze™ HFC, Revco™ CxF, TSC Series
Monitoring Options Alarm, Recorder
Voltage 0/5 V