Learn More
Fisherbrand™ Traceable™ Data Acquisition Software
Catalog No. 03391121
Change view
Click to view available options
Includes:
Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
03391121 Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
03391124 Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03391121 Supplier Fisherbrand™ Supplier No. 3780413
Complete Data Acquisition Software enables measurement readings to be displayed and stored on a PC from up to 8 devices at a time.
  • Display and store measurement readings on a PC from up to 8 devices at a time
  • Data capture runs in the background, enabling users to work on other programs simultaneously
  • Set audible and visual notification alarms, or immediately send an email if an over/under condition occurs
  • Software can be used to connect a meter directly to PC via cable
  • Check software specification tables for meter compatibility
Specifications
Type Data Acquisition Software
Display Type No Display
Description Data Acquisition Software with USB Cable
Includes Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
For Use With (Equipment) 37803-92, 37803-99, 91210-03, 19601-08 (Traceable™ 4132, 4310, 4137, 4169)