Learn More
STERIS Dart™ Daily Air Removal Test
Catalog No. 296362144
Change view
Click to view available options
Quantity:
5 Pk./Cs. (10 Tests/Pk.)
Catalog No. Quantity
296362144 5 Pk./Cs. (10 Tests/Pk.)
1 options
Catalog No. 296362144 Supplier STERIS Supplier No. NB113
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Dart™ Daily Air Removal Test is used in monitoring healthcare steam sterilizers.
Features:
  • Disposable
  • Preassembled Single Use test
  • Functions as a AAMI Bowie Dick type test
  • Light weight and compact
  • Indicator strip can be kept as a permanent record
  • Lot number and expiration date on each test
Benefits:
  • Eliminates the need to construct test packs while providing consistent quality
  • Non-uniform color change provides an immediate indication of inadequate air removal from the sterilizer chamber during the cycle
  • Eliminates the need for bulky towels or wrappers
  • Documentation of the efficacy of the sterilizer to record removed air from the pack
  • Traceability and recognition of expired packs at time of use
  • Used in monitoring healthcare steam sterilizers.
  • Meets ANSI/AAMI/ISO 11140-4 requirements for Type 2 Air Removal Indicator Systems and operates at 270 to 274°F/132 to 134°C with an exposure time of 3.5 to 4.0 minutes.
  • Easy to use.
  • Users can eliminate waste associated with paper box products.
  • Compact barrel design creates a barrier to air removal and steam penetration.
  • Special record forms are included for convenience
User Process:
  • Placed directly into an otherwise empty steam sterilizer chamber and does not require a retaining device.
  • Optional tie is provided for suspension of the DART test.
  • During processing, steam must displace the air through the blue heat sink and then within the restricted transparent chamber of the chemical indicator.
  • Results are immediately visible through the clear vial.
  • A uniform color change from yellow to black indicates that all the air was removed and replaced by steam.
  • When the test is completed, simply snap off the window, slide out the chemical indicator, and record the results.
Specifications
Certifications/Compliance ANSI/AAMI/ISO 11140-4 requirements for Type 2 Air Removal Indicator Systems
Product Type Disposable Preassembled Single Use Test
Quantity 5 Pk./Cs. (10 Tests/Pk.)