missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ DAR-4M AM, Calbiochem™,
Specifications
Specifications
| Color | Red |
| Quantity | 1 mg |
| Formula Weight | 630.5 |
| Grade | HPLC |
| Physical Form | Liquid |
| Chemical Name or Material | DAR-4M AM |
Safety and Handling
| Recommended Storage | -20°C (-4°F) |