Learn More
Selleck Chemical Llc Daporinad (FK866, APO866) 5mg 658084-64-1 DFS Item
Supplier:  Selleck Chemicals Co Ltd S27995MG
Daporinad (FK866, APO866) effectively inhibits nicotinamide phosphoribosyltransferase (NMPRTase; Nampt) with IC50 of 0.09 nM in a cell-free assay. Daporinad (FK866, APO866) triggers autophagy. Phase 1/2. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
Catalog No. 501362977
Only null left
Explore available promotions