missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ DAPI, Dihydrochloride, Calbiochem™,

Catalog No. 26829810mg
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26829810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26829810MG Supplier MilliporeSigma™ Supplier No. 26829810MG
Only null left

Cell-permeable DNA-binding dye

Chemical Identifiers

CAS 28718-90-3
Molecular Formula C16H17Cl2N5
Molecular Weight (g/mol) 350.25
MDL Number MFCD00012681
InChI Key FPNZBYLXNYPRLR-UHFFFAOYSA-N
Synonym 4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836
PubChem CID 160166
IUPAC Name dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N

Specifications

Color Yellow
Quantity 10 mg
Solubility Information Easily soluble in cold water
Formula Weight 350.3
Percent Purity ≥90%
Physical Form Solid
Chemical Name or Material DAPI, Dihydrochloride
RTECSNumber NL5995050
Recommended Storage -20°C (-4°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).