missing translation for 'onlineSavingsMsg'
Learn More

Dantrolene sodium salt

Catalog No. AAJ60887MD
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60887MD 250 mg
AAJ60887MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60887MD Supplier Thermo Scientific Chemicals Supplier No. J60887MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibits intracellular calcium release from the sarcoplasmic reticulum | CAS: 14663-23-1 | C14H12N4NaO5 | 339.263 g/mol

Inhibits intracellular calcium release from the sarcoplasmic reticulum

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14663-23-1
Molecular Formula C14H12N4NaO5
Molecular Weight (g/mol) 339.263
MDL Number MFCD00079130
InChI Key XHONPQLVCHTEDY-YVCISUPJSA-N
PubChem CID 131674048
IUPAC Name molecular hydrogen;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione;sodium
SMILES [HH].C1C(=O)NC(=O)N1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+](=O)[O-].[Na]

Specifications

Color Orange to Yellow
Quantity 250 mg
Merck Index 14,2816
Formula Weight 336.23
Physical Form Crystalline Powder
Chemical Name or Material Dantrolene sodium salt
RTECSNumber MU3875000

RUO – Research Use Only