missing translation for 'onlineSavingsMsg'
Learn More

Dantrolene Sodium Salt Hydrate 98.0+%, TCI America™

Catalog No. p-7128039
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39961G 1 g
D39965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39961G Supplier TCI America Supplier No. D39961G
Only null left

Chemical Identifiers

CAS 14663-23-1
Molecular Formula C14H12N4NaO5
Molecular Weight (g/mol) 339.263
MDL Number MFCD00079130
InChI Key XHONPQLVCHTEDY-YVCISUPJSA-N
Synonym 1-[5-(4-Nitrophenyl)furfurylideneamino]hydantoin Sodium Salt
PubChem CID 131674048
IUPAC Name molecular hydrogen;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione;sodium
SMILES [HH].C1C(=O)NC(=O)N1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+](=O)[O-].[Na]

Specifications

Quantity 1 g
Formula Weight 336.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dantrolene Sodium Salt Hydrate
RTECSNumber MU3875000
TSCA No