Learn More
New England Biolab Ltd dam-/dcm- Competent E. coli 6 x 0.2 ml/tube
Supplier: New England Biolab Ltd C2925I
Methyltransferase deficient chemically competent E. coli cells suitable for growth of plasmids free of Dam and Dcm methylation. Note that dam- strains are not recommended as a host for primary cloning/ligation. The dam mutation can result in an increased mutation rate in the cell and a reduction in the transformation efficiency. DNA should be maintained in a dam+ strain unless there is a specific need for DNA free of Dam or Dcm methylation.
Description
- Environmental benefits include:
- Renewable Energy
- Free-of a substance that causes environmental harm
- Ozone-Safe or Safe for the Ozone Layer
- Non-Toxic