missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daidzin, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 460420010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Daidzin | |
| 2.5% max. | |
| 97.5% min. (HPLC) | |
| C21H20O9 | |
| 1g | |
| KYQZWONCHDNPDP-QNDFHXLGSA-N | |
| 3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one | |
| 107971 | |
| 416.38 |
| 552-66-9 | |
| Authentic | |
| Glass Bottle | |
| MFCD00017466 | |
| daidzin, daidzoside, daidzein 7-o-glucoside, daidzein 7-glucoside, daidzein-7-glucoside, daidzein 7-o-beta-d-glucoside, unii-4r2x91a5m5, daidzein-7-o-glucoside, 7-beta-d-glucopyranosyloxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 7-o-b-d-glucopyranoside | |
| OC[C@H]1O[C@@H](OC2=CC=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O | |
| 416.38 | |
| CHEBI:42202 | |
| 98% |
Chemical Identifiers
| 552-66-9 | |
| 416.38 | |
| KYQZWONCHDNPDP-QNDFHXLGSA-N | |
| 107971 | |
| 3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one |
| C21H20O9 | |
| MFCD00017466 | |
| daidzin, daidzoside, daidzein 7-o-glucoside, daidzein 7-glucoside, daidzein-7-glucoside, daidzein 7-o-beta-d-glucoside, unii-4r2x91a5m5, daidzein-7-o-glucoside, 7-beta-d-glucopyranosyloxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 7-o-b-d-glucopyranoside | |
| CHEBI:42202 | |
| OC[C@H]1O[C@@H](OC2=CC=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O |
RUO â Research Use Only