missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ DAF-2 DA, Calbiochem™,

Catalog No. 2515051MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2515051MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2515051MG Supplier MilliporeSigma™ Supplier No. 2515051MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell-permeable derivative of DAF-2 that is hydrolyzed to DAF-2 by intracellular esterases

Chemical Identifiers

CAS 205391-02-2
Molecular Formula C24H18N2O7
Molecular Weight (g/mol) 446.42
MDL Number MFCD05865101
InChI Key PTSUYDXEEKDBQU-UHFFFAOYSA-N
Synonym daf-2 da, 4,5-diaminofluorescein diacetate, 5,6-diaminofluorescein diacetate, daf-2 da cell permeable, daf 2da, daf-2da, 5,6-diaminofluorescein diacetat, 4,5-diaminofluorescein diacetate hplc, daf-2 da solution 5 mm in dmso , 1 mg in 0.45 ml dmso, 4,5-diaminofluorescein diacetate/ 5,6-diaminofluorescein diacetate
PubChem CID 6603693
IUPAC Name 3'-(acetyloxy)-5,6-diamino-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-6'-yl acetate
SMILES CC(=O)OC1=CC2=C(C=C1)C1(OC(=O)C3=CC(N)=C(N)C=C13)C1=C(O2)C=C(OC(C)=O)C=C1

Specifications

Color Yellow
Quantity 1 mg
Formula Weight 446.4
Percent Purity ≥98%
Grade HPLC
Concentration or Composition (by Analyte or Components) 5mM solution in DMSO
Physical Form Liquid
Chemical Name or Material DAF-2 DA
Recommended Storage -20°C (-4°F); Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.