missing translation for 'onlineSavingsMsg'
Learn More

Dacarbazine

Catalog No. p-7042006
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61023ME 500 mg
AAJ6102303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61023ME Supplier Thermo Scientific Chemicals Supplier No. J61023ME
Only null left

CAS: 4342-03-4 | C6H10N6O | 182.19 g/mol

Used in treatment of malignant melanoma and sarcomas. Generates methyl adducts to purine bases in DNA

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used in treatment of malignant melanoma and sarcomas
  • Generates methyl adducts to purine bases in DNA

Chemical Identifiers

CAS 3-4-4342
MDL Number MFCD00057167

Specifications

Melting Point 210°C to 212°C
Color White to Yellow
Molecular Formula C4H3IN2O
Quantity 500 mg
Merck Index 14,2798
Synonym 5-iodopyrimidin-4-ol, 5-iodopyrimidin-4 3h-one, 4-hydroxy-5-iodopyrimidine, 5-iodo-1h-pyrimidin-4-one, 4 1h-pyrimidinone, 5-iodo, 5-iodopyrimidin-4 1h-one, 5-iodo-3h-pyrimidin-4-one, 5-iodo-3,4-dihydropyrimidin-4-one, 5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one
Solubility Information Slightly soluble in water,DMSO and ethanol /n
InChI Key SMIGOGPUTQXOKF-UHFFFAOYSA-N
SMILES C1=C(C(=O)NC=N1)I
IUPAC Name 5-iodo-1H-pyrimidin-6-one
Molecular Weight (g/mol) 221.99
PubChem CID 12356582
Formula Weight 182.18
Physical Form Powder
Chemical Name or Material Dacarbazine
Show More Show Less
Hazard Category H312+H332-H315-H319-H335-H350
Hazard Statement GHS H Statement
H311-H340-H350-H332-H315-H319-H335
Toxic in contact with skin.
May cause genetic defects.
May cause cancer.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P332+P313-P362-P501c
EINECSNumber 224-396-1
RTECSNumber NI3950000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only